Decamethylcyclopentasiloxane ≥99.0%
Supplier: TCI America
D1890-500ML
D1890-25ML
D1890-100ML
TCD1890-500MLEA
314.42
CAD
TCD1890-500ML
TCD1890-025ML
TCD1890-100ML
Decamethylcyclopentasiloxane ≥99.0%
Decamethylcyclopentasiloxane
CAS Number: 541-02-6
MDL Number: MFCD00046966
Molecular Formula: C10H30O5Si5
Molecular Weight: 370.77
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 210
Melting point (°C): -38
Flash Point (°C): 76
Specific Gravity (20/20): 0.96
Formula:
C₁₀H₃₀O₅Si₅ Boiling Pt: 210 °C Melting Pt: –38 °C Density: 0.96 (20 °C) Flash Pt: 76 °C Storage Temperature: Ambient |
MDL Number:
MFCD00046966 CAS Number: 541-02-6 EINECS: 208-764-9 Merck Index: 12,02905 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...