(3,4-Dimethoxyphenyl)acetonitrile ≥98.0%
Supplier: TCI America
Synonyms:
3,4-Dimethoxyphenylacetonitrile, Homoveratronitrile, 3,4-Dimethoxybenzyl cyanide
D1887-500G
D1887-25G
TCD1887-500GEA
301.78
CAD
TCD1887-500G
TCD1887-025G
(3,4-Dimethoxyphenyl)acetonitrile ≥98.0%
(3,4-Dimethoxyphenyl)acetonitrile
CAS Number: 93-17-4
MDL Number: MFCD00001911
Molecular Formula: C10H11NO2
Molecular Weight: 177.20
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 160
Melting point (°C): 65
Flash Point (°C): 250
Other Product Names / Synonyms: 3,4-Dimethoxyphenylacetonitrile, Homoveratronitrile
Formula:
C₁₀H₁₁NO₂ MW: 177.2 g/mol Boiling Pt: 160 °C Melting Pt: 65 °C Flash Pt: 250 °C |
MDL Number:
MFCD00001911 CAS Number: 93-17-4 EINECS: 202-225-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...