Ethyl diethoxyacetate ≥97.0%
Supplier: TCI America
D1883-25ML
D1883-500ML
D1883-100ML
TCD1883-025MLEA
147.1
CAD
TCD1883-025ML
TCD1883-500ML
TCD1883-100ML
Ethyl diethoxyacetate ≥97.0%
Ethyl diethoxyacetate
CAS Number: 6065-82-3
MDL Number: MFCD00009231
Molecular Formula: C8H16O4
Molecular Weight: 176.21
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 84
Flash Point (°C): 85
Specific Gravity (20/20): 0.99
Other Product Names / Synonyms: Diethoxyacetic Acid Ethyl Ester, Ethyl Glyoxylate Diethyl Acetal, Glyoxylic Acid Ethyl Ester Diethyl Acetal
Formula:
C₈H₁₆O₄ Boiling Pt: 84 °C Density: 0.99 (20 °C) Flash Pt: 85 °C |
MDL Number:
MFCD00009231 CAS Number: 6065-82-3 EINECS: 227-999-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...