2',5'-Dimethylacetophenone ≥98.0%

Supplier: TCI America

D1880-250ML D1880-25ML
TCD1880-250MLEA 555.37 CAD
TCD1880-250ML TCD1880-025ML
2',5'-Dimethylacetophenone ≥98.0%
2',5'-Dimethylacetophenone

CAS Number: 2142-73-6
MDL Number: MFCD00014988
Molecular Formula: C10H12O
Molecular Weight: 148.21
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 112
Flash Point (°C): 96
Specific Gravity (20/20): 1.00


Other Product Names / Synonyms: 2-Acetyl-p-xylene

Formula: C₁₀H₁₂O
Boiling Pt: 112 °C
Melting Pt: –18 °C
Density: 1.00 (20 °C)
Flash Pt: 96 °C
MDL Number: MFCD00014988
CAS Number: 2142-73-6
EINECS: 218-399-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a8971d7133507be37eab8f84f2a11f279050a76&vid=8a8971d7133507be37eab8f84f2a11f279050a76&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR