2',4'-Dimethylacetophenone ≥95.0%
Supplier: TCI America
D1879-5ML
D1879-25ML
TCD1879-5MLEA
53.72
CAD
TCD1879-5ML
TCD1879-025ML
2',4'-Dimethylacetophenone ≥95.0%
2',4'-Dimethylacetophenone
CAS Number: 89-74-7
MDL Number: MFCD00003571
Molecular Formula: C10H12O
Molecular Weight: 148.21
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 122
Flash Point (°C): 100
Specific Gravity (20/20): 1.00
Other Product Names / Synonyms: 4-Acetyl-m-xylene
Formula:
C₁₀H₁₂O Boiling Pt: 122 °C Density: 1.00 (20 °C) Flash Pt: 100 °C |
MDL Number:
MFCD00003571 CAS Number: 89-74-7 EINECS: 201-935-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...