2,5-Dichloro-p-phenylenediamine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
D1873-250G
D1873-25G
TCD1873-250GEA
251.22
CAD
TCD1873-250G
TCD1873-025G
2,5-Dichloro-p-phenylenediamine ≥98.0% (by GC, titration analysis)
2,5-Dichloro-p-phenylenediamine
CAS Number: 20103-09-7
MDL Number: MFCD00007902
Molecular Formula: C6H6Cl2N2
Molecular Weight: 177.03
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 166
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 1,4-Diamino-2,5-dichlorobenzene
Formula:
C₆H₆Cl₂N₂ Melting Pt: 166 °C |
MDL Number:
MFCD00007902 CAS Number: 20103-09-7 EINECS: 243-512-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...