2,6-Dichloropyrazine ≥99.0%

Supplier: TCI America
Warning

D1867-5G D1867-25G
TCD1867-005GEA 46.29 CAD
TCD1867-005G TCD1867-025G
2,6-Dichloropyrazine ≥99.0%
2,6-Dichloropyrazine

CAS Number: 4774-14-5
MDL Number: MFCD00006125
Molecular Formula: C4H2Cl2N2
Molecular Weight: 148.97
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 122
Melting point (°C): 54
Flash Point (°C): 110

Formula: C₄H₂Cl₂N₂
Boiling Pt: 122 °C
Melting Pt: 54 °C
Density: 1.713 g/cm³ (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00006125
CAS Number: 4774-14-5
EINECS: 225-316-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8338b23f3d14fb90fc280984122d54fc610acf8b&vid=8338b23f3d14fb90fc280984122d54fc610acf8b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR