1,4-Dibromo-2,5-difluorobenzene ≥98.0%

Supplier: TCI America
Warning

D1864-25G D1864-10G
TCD1864-25GEA 184.86 CAD
TCD1864-25G TCD1864-010G
1,4-Dibromo-2,5-difluorobenzene ≥98.0%
1,4-Dibromo-2,5-difluorobenzene

CAS Number: 327-51-5
MDL Number: MFCD00000346
Molecular Formula: C6H2Br2F2
Molecular Weight: 271.89
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 96
Melting point (°C): 65

Formula: C₆H₂Br₂F₂
Boiling Pt: 96 °C
Melting Pt: 65 °C
MDL Number: MFCD00000346
CAS Number: 327-51-5
EINECS: 206-317-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6953d457c66a6de9c766bd26ae2ffa3be6de2c9&vid=c6114ced5fc8f88ff150ac3057e9b8466fd14fe2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR