2,4-Dibromoanisole ≥98.0%
Supplier: TCI America
|
Warning
|
Synonyms:
1,3-Dibromo-4-methoxybenzene, 2,4-Dibromo-1-methoxybenzene
D1863-250G
D1863-25G
TCD1863-250GEA
884.8
CAD
TCD1863-250G
TCD1863-025G
2,4-Dibromoanisole ≥98.0%
2,4-Dibromoanisole
CAS Number: 21702-84-1
MDL Number: MFCD00000079
Molecular Formula: C7H6Br2O
Molecular Weight: 265.93
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 101
Melting point (°C): 63
Formula:
C₇H₆Br₂O Boiling Pt: 101 °C Melting Pt: 63 °C |
MDL Number:
MFCD00000079 CAS Number: 21702-84-1 EINECS: 244-536-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...