2,5-Dibromo-p-xylene ≥98.0%
Supplier: TCI America
D1841-25G
D1841-100G
TCD1841-025GEA
242.42
CAD
TCD1841-025G
76218-986
2,5-Dibromo-p-xylene ≥98.0%
2,5-Dibromo-p-xylene
CAS Number: 1074-24-4
MDL Number: MFCD00000091
Molecular Formula: C8H8Br2
Molecular Weight: 263.96
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 150
Melting point (°C): 75
Flash Point (°C): 110
Other Product Names / Synonyms: 1,4-Dibromo-2,5-dimethylbenzene
Formula:
C₈H₈Br₂ Boiling Pt: 150 °C Melting Pt: 75 °C Flash Pt: 110 °C |
MDL Number:
MFCD00000091 CAS Number: 1074-24-4 EINECS: 214-038-2 UN: 3077 ADR: 9,III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...