2,4-Difluorobenzoic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

D1833-25G D1833-500G
TCD1833-025GEA 63.2 CAD
TCD1833-025G TCD1833-500G
2,4-Difluorobenzoic acid ≥98.0% (by titrimetric analysis)
2,4-Difluorobenzoic acid

CAS Number: 1583-58-0
MDL Number: MFCD00011670
Molecular Formula: C7H4F2O2
Molecular Weight: 158.10
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 188

Formula: C₇H₄F₂O₂
Boiling Pt: 241 °C (1013 hPa)
Melting Pt: 188 °C
Storage Temperature: Ambient
MDL Number: MFCD00011670
CAS Number: 1583-58-0
EINECS: 216-430-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=21cb89baee89b0642dd4bc1a6c56fd84d654c000&vid=5bf46d57ecd29bdb56ce95d87cffea782b1372ea&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR