4,6-Dihydroxypyrimidine ≥95.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 6-Hydroxy-1H-pyrimidin-4-one

D1821-250G D1821-25G
TCD1821-250GEA 287.56 CAD
TCD1821-250G TCD1821-025G
4,6-Dihydroxypyrimidine ≥95.0% (by titrimetric analysis)
4,6-Dihydroxypyrimidine

CAS Number: 1193-24-4
MDL Number: MFCD00016733
Molecular Formula: C4H4N2O2
Molecular Weight: 112.09
Purity/Analysis Method: >95.0% (T)
Form: Crystal


Other Product Names / Synonyms: 4,6-Pyrimidinediol

Formula: C₄H₄N₂O₂
MW: 112.09 g/mol
Melting Pt: > 300 °C
MDL Number: MFCD00016733
CAS Number: 1193-24-4
EINECS: 214-772-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f71e2150c78cda1d1df7dbe5730a5c71ebab531&vid=6f71e2150c78cda1d1df7dbe5730a5c71ebab531&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR