(3,4-Dichlorophenyl)acetic acid ≥98.0%

Supplier: TCI America

Synonyms: 3,4-Dichlorophenylacetic acid

D1813-25G D1813-500G
TCD1813-025GEA 66.36 CAD
TCD1813-025G TCD1813-500G
(3,4-Dichlorophenyl)acetic acid ≥98.0%
(3,4-Dichlorophenyl)acetic acid

CAS Number: 5807-30-7
MDL Number: MFCD00004333
Molecular Formula: C8H6Cl2O2
Molecular Weight: 205.03
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 90

Formula: (C₆H₃Cl₂)CH₂COOH
MW: 205.04 g/mol
Boiling Pt: 314
Melting Pt: 90 °C
MDL Number: MFCD00004333
CAS Number: 5807-30-7
EINECS: 227-368-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=110ead11933bbc93d83adc958fb9713bc25c8dcb&vid=110ead11933bbc93d83adc958fb9713bc25c8dcb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR