2,3-Dichlorotoluene ≥98.0%

Supplier: TCI America

D1805-25G D1805-500G
TCD1805-025GEA 85.32 CAD
TCD1805-025G TCD1805-500G
2,3-Dichlorotoluene ≥98.0%
2,3-Dichlorotoluene

CAS Number: 32768-54-0
MDL Number: MFCD00000546
Molecular Formula: C7H6Cl2
Molecular Weight: 161.03
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 208
Flash Point (°C): 83
Freezing point (°C): 4
Specific Gravity (20/20): 1.27

Formula: C₇H₆Cl₂
Boiling Pt: 208 °C
Melting Pt: 6 °C
Density: 1.27 (20 °C)
Flash Pt: 83 °C
MDL Number: MFCD00000546
CAS Number: 32768-54-0
EINECS: 251-203-8
UN: 3082
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=55bea58dc1c7fac49611a9734378b560d2811efa&vid=7c8ec16919b2e14e3e2de4abc7b12a7e77112ad9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR