4,4'-Difluorobenzophenone ≥99.0%

Supplier: TCI America
Warning

Synonyms: Bis(4-Fluorophenyl)methanone, Bis(4-fluorophenyl) ketone

D1797-25G D1797-500G D1797-100G
TCD1797-025GEA 50.56 CAD
TCD1797-025G TCD1797-500G TCD1797-100G
4,4'-Difluorobenzophenone ≥99.0%
4,4'-Difluorobenzophenone

CAS Number: 345-92-6
MDL Number: MFCD00000353
Molecular Formula: C13H8F2O
Molecular Weight: 218.20
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 108

Formula: C₁₃H₈F₂O
MW: 218.2 g/mol
Boiling Pt: 170…172 °C (10 mmHg)
Melting Pt: 108 °C
Density: 1.39 g/cm³ (20 °C)
MDL Number: MFCD00000353
CAS Number: 345-92-6
EINECS: 206-466-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7a289d5230f554dfb16215bf76480614f073d2af&vid=7a289d5230f554dfb16215bf76480614f073d2af&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR