2,2'-Dinitrobiphenyl ≥99.0%

Supplier: TCI America

D1777-5G D1777-25G
TCD1777-005GEA 178.54 CAD
TCD1777-005G TCD1777-025G
2,2'-Dinitrobiphenyl ≥99.0%
2,2'-Dinitrobiphenyl

CAS Number: 2436-96-6
MDL Number: MFCD00007127
Molecular Formula: C12H8N2O4
Molecular Weight: 244.21
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 125

Formula: C₁₂H₈N₂O₄
Boiling Pt: 305 °C (1013 hPa)
Melting Pt: 125 °C
Density: 1.45 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007127
CAS Number: 2436-96-6
EINECS: 219-435-4
UN: 3143
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd8b49ca1bfd7a878ba48dcfa6129d59d95a7249&vid=fd8b49ca1bfd7a878ba48dcfa6129d59d95a7249&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR