Sinapic acid ≥98.0%

Supplier: TCI America
Warning

Synonyms: 3,5-Dimethoxy-4-hydroxycinnamic acid, 3',5'-Dimethoxy-4'-hydroxycinnamic acid, 4-Hydroxy-3,5-dimethoxycinnamic acid

D1765-5G D1765-25G
TCD1765-005GEA 135.67 CAD
TCD1765-005G TCD1765-025G
Sinapic acid ≥98.0%
Sinapic acid

CAS Number: 530-59-6
MDL Number: MFCD00004401
Molecular Formula: C11H12O5
Molecular Weight: 224.21
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 198


Product Categories: Bioscience, Phenylpropanoids, Phenylpropanoids (Phytochemicals)
Other Product Names / Synonyms: 4-Hydroxy-3,5-dimethoxycinnamic Acid, Sinapinic Acid

Formula: C₁₁H₁₂O₅
Boiling Pt: 378 °C (1013 hPa)
Melting Pt: 198 °C
MDL Number: MFCD00004401
CAS Number: 530-59-6
EINECS: 208-487-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c39cd23bc059909474aac093fb93020b5e1fa35c&vid=c39cd23bc059909474aac093fb93020b5e1fa35c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR