1,4-Dithiane-2,5-diol ≥96.0%

Supplier: TCI America

D1761-25G D1761-500G
TCD1761-025GEA 77.42 CAD
TCD1761-025G TCD1761-500G
1,4-Dithiane-2,5-diol ≥96.0%
1,4-Dithiane-2,5-diol

CAS Number: 40018-26-6
MDL Number: MFCD00006659
Molecular Formula: C4H8O2S2
Molecular Weight: 152.23
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 1,4-Dithiane-2,5-diol, Mercaptoacetaldehyde Dimer

Formula: C₄H₈O₂S₂
MDL Number: MFCD00006659
CAS Number: 40018-26-6
EINECS: 254-751-6
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7e43dcb51379995973c72db4c66a8bd37e253447&vid=7e43dcb51379995973c72db4c66a8bd37e253447&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR