2,6-Diisopropylaniline ≥90.0%
Supplier: TCI America
D1755-500ML
D1755-25ML
D1755-100ML
TCD1755-500MLEA
712.58
CAD
TCD1755-500ML
TCD1755-025ML
TCD1755-100ML
2,6-Diisopropylaniline ≥90.0%
2,6-Diisopropylaniline
CAS Number: 24544-04-5
MDL Number: MFCD00008887
Molecular Formula: C12H19N
Molecular Weight: 177.29
Purity/Analysis Method: >90.0% (GC)
Form: Clear Liquid
Boiling point (°C): 122
Melting point (°C): -45
Flash Point (°C): 117
Specific Gravity (20/20): 0.94
Formula:
C₁₂H₁₉N MW: 177.29 g/mol Boiling Pt: 257 Melting Pt: -45 Density: 0.94 (20 °C) Flash Pt: 117 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008887 CAS Number: 24544-04-5 EINECS: 246-305-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...