Ethyl-4-(dimethylamino)benzoate ≥98.0%

Supplier: TCI America

Synonyms: 4-(Dimethylamino)benzoic acid ethyl ester, Ethyl 4-dimethylaminobenzoate

D1744-500G D1744-25G D1744-100G
TCD1744-500GEA 323.9 CAD
TCD1744-500G TCD1744-025G TCD1744-100G
Ethyl-4-(dimethylamino)benzoate ≥98.0%
Ethyl 4-(dimethylamino)benzoate

CAS Number: 10287-53-3
MDL Number: MFCD00009115
Molecular Formula: C11H15NO2
Molecular Weight: 193.25
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 191
Melting point (°C): 66
Flash Point (°C): 200


Other Product Names / Synonyms: 4-(Dimethylamino)benzoic Acid Ethyl Ester

Formula: C₁₁H₁₅NO₂
MW: 193.25 g/mol
Boiling Pt: 112
Melting Pt: 66 °C
Density: 1.0099 g/cm³ (99.7 °C)
Flash Pt: 200 °C
MDL Number: MFCD00009115
CAS Number: 10287-53-3
EINECS: 233-634-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=23ecd96d9ef54f45aae1f046a1bb76e6985e39ce&vid=23ecd96d9ef54f45aae1f046a1bb76e6985e39ce&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR