Dimethoxy(diphenyl)silane ≥98.0%

Supplier: TCI America

D1731-500ML D1731-25ML D1731-100ML
TCD1731-500MLEA 359.94 CAD
TCD1731-500ML TCD1731-025ML TCD1731-100ML
Dimethoxy(diphenyl)silane ≥98.0%
Dimethoxy(diphenyl)silane

CAS Number: 6843-66-9
MDL Number: MFCD00025690
Molecular Formula: C14H16O2Si
Molecular Weight: 244.37
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 110
Flash Point (°C): 157
Specific Gravity (20/20): 1.08

Formula: C₁₄H₁₆O₂Si
Boiling Pt: 110 °C
Density: 1.08 (20 °C)
Flash Pt: 157 °C
MDL Number: MFCD00025690
CAS Number: 6843-66-9
EINECS: 229-929-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=be0d2070a6ef51ea5addee94fa5cef5302e6dc3c&vid=be0d2070a6ef51ea5addee94fa5cef5302e6dc3c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR