(R)-(-)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol ≥97.0%
Supplier: TCI America
Synonyms:
(R)-(-)-1,2-O-Isopropylidenglycerol, (R)-(-)-2,2-Dimethyl-1,3-dioxolane-4-methanol, (R)-(-)-2,3-O-Isopropylideneglycerol, (R)-(-)-1,2-O-Isopropylideneglycerol, (R)-(2,2-Dimethyl-1,3-dioxolan-4-yl)methanol
CAS Number: 14347-78-5
MDL Number: MFCD00003213
Molecular Formula: C6H12O3
Molecular Weight: 132.16
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 78
Flash Point (°C): 80
Specific Gravity (20/20): 1.07
Specific rotation [a]20/D: -14 deg (neat)
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: L-(-)-1,2-Isopropylideneglycerol
Formula:
C₆H₁₂O₃ Boiling Pt: 78 °C Density: 1.07 (20 °C) Flash Pt: 80 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00003213 CAS Number: 14347-78-5 EINECS: 405-320-8 Merck Index: 12,05232 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...