9,10-Diphenylanthracene ≥98.0%
Supplier: TCI America
D1689-1G
D1689-25G
TCD1689-001GEA
66.36
CAD
TCD1689-001G
TCD1689-025G
9,10-Diphenylanthracene ≥98.0%
9,10-Diphenylanthracene
CAS Number: 1499-10-1
MDL Number: MFCD00001253
Molecular Formula: C26H18
Molecular Weight: 330.43
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 485
Lambda max.: 393 nm (Cyclohexane)
Formula:
C₂₆H₁₈ Boiling Pt: 485 °C Melting Pt: 245 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001253 CAS Number: 1499-10-1 EINECS: 216-105-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...