3,3'-Dinitrobenzophenone ≥99.0%

Supplier: TCI America

D1688-5G D1688-25G
TCD1688-005GEA 491.11 CAD
TCD1688-005G TCD1688-025G
3,3'-Dinitrobenzophenone ≥99.0%
3,3'-Dinitrobenzophenone

CAS Number: 21222-05-9
MDL Number: MFCD00039740
Molecular Formula: C13H8N2O5
Molecular Weight: 272.22
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 156

Formula: C₁₃H₈N₂O₅
Melting Pt: 156 °C
MDL Number: MFCD00039740
CAS Number: 21222-05-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=afe8482d563e7e8edffc8d0d70d822b5642a9350&vid=afe8482d563e7e8edffc8d0d70d822b5642a9350&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR