3,3'-Methylenedianiline ≥98.0%

Supplier: TCI America

Synonyms: 3,3'-Methylenedianiline, 3,3'-Diaminodiphenylmethane

D1683-5G D1683-1G
TCD1683-005GEA 747.34 CAD
TCD1683-005G TCD1683-1G
3,3'-Methylenedianiline ≥98.0%
3,3'-Methylenedianiline

CAS Number: 19471-12-6
MDL Number: MFCD00014777
Molecular Formula: C13H14N2
Molecular Weight: 198.27
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 85


Product Categories: Materials Chemistry, Monomers, Polyimide Raw Materials, Diphenylmethanes (for High-Performance Polymer Research)
Other Product Names / Synonyms: 3,3'-Methylenedianiline

Formula: C₁₃H₁₄N₂
Melting Pt: 85 °C
MDL Number: MFCD00014777
CAS Number: 19471-12-6
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aa25fb1e176f38819c5a1e0bd61937018c985f6e&vid=aa25fb1e176f38819c5a1e0bd61937018c985f6e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR