3,3'-Diaminobenzophenone ≥95.0% (by GC, titration analysis)
Supplier: TCI America
D1682-5G
D1682-1G
TCD1682-005GEA
656.17
CAD
TCD1682-005G
TCD1682-001G
3,3'-Diaminobenzophenone ≥95.0% (by GC, titration analysis)
3,3'-Diaminobenzophenone
CAS Number: 611-79-0
MDL Number: MFCD00014774
Molecular Formula: C13H12N2O
Molecular Weight: 212.25
Purity/Analysis Method: >95.0% (GC,T)
Form: Crystal
Melting point (°C): 153
Formula:
C₁₃H₁₂N₂O Melting Pt: 153 °C |
MDL Number:
MFCD00014774 CAS Number: 611-79-0 EINECS: 210-281-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...