2-(Dimethylamino)pyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

D1680-500G D1680-25G
TCD1680-500GEA 1082.3 CAD
TCD1680-500G TCD1680-25G
2-(Dimethylamino)pyridine ≥98.0% (by GC, titration analysis)
2-(Dimethylamino)pyridine

CAS Number: 5683-33-0
MDL Number: MFCD00006261
Molecular Formula: C7H10N2
Molecular Weight: 122.17
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 108
Flash Point (°C): 74
Specific Gravity (20/20): 1.01


Other Product Names / Synonyms: N-(2-Pyridyl)dimethylamine

Formula: C₇H₁₀N₂
Boiling Pt: 108 °C
Density: 1.01 (20 °C)
Flash Pt: 74 °C
Storage Temperature: Ambient
MDL Number: MFCD00006261
CAS Number: 5683-33-0
EINECS: 227-147-5
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a283b089b14ed77f56d57fc2da709353bd78e46b&vid=a283b089b14ed77f56d57fc2da709353bd78e46b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR