2,2,6,6-Tetramethyl-3,5-heptanedione ≥97.0%
Supplier: TCI America
D1678-5G
D1678-25G
TCD1678-005GEA
88.48
CAD
TCD1678-005G
TCD1678-025G
2,2,6,6-Tetramethyl-3,5-heptanedione ≥97.0%
2,2,6,6-Tetramethyl-3,5-heptanedione
CAS Number: 1118-71-4
MDL Number: MFCD00008848
Molecular Formula: C11H20O2
Molecular Weight: 184.28
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 80
Flash Point (°C): 67
Freezing point (°C): 19
Specific Gravity (20/20): 0.90
Product Categories: Synthetic Organic Chemistry, Oxidation, Ligands
Other Product Names / Synonyms: 2,2,6,6-Tetramethyl-3,5-heptanedione
Formula:
C₁₁H₂₀O₂ Boiling Pt: 80 °C Melting Pt: 28…30 °C Density: 0.90 (20 °C) Flash Pt: 67 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008848 CAS Number: 1118-71-4 EINECS: 214-268-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...