2,4-Dimethoxybenzoic acid ≥99.0%

Supplier: TCI America

D1675-25G D1675-5G
TCD1675-025GEA 69.52 CAD
TCD1675-025G 76218-976
2,4-Dimethoxybenzoic acid ≥99.0%
2,4-Dimethoxybenzoic acid

CAS Number: 91-52-1
MDL Number: MFCD00002434
Molecular Formula: C9H10O4
Molecular Weight: 182.18
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 110

Formula: C₉H₁₀O₄
MW: 182.18 g/mol
Boiling Pt: 313
Melting Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00002434
CAS Number: 91-52-1
EINECS: 202-074-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1c2c65ac95036b29c611ba218e3656a1c26e51da&vid=1c2c65ac95036b29c611ba218e3656a1c26e51da&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR