2-Chloropropyldimethylammonium chloride ≥98.0%

Supplier: TCI America

Synonyms: N-(2-Chloropropyl)-N,N-dimethylammonium chloride, 2-Chloro-N,N-dimethylpropan-1-amine hydrochloride, 2-Dimethylaminoisopropyl chloride hydrochloride

D1665-25G D1665-500G
TCD1665-025GEA 45.82 CAD
TCD1665-025G TCD1665-500G
2-Chloropropyldimethylammonium chloride ≥98.0%
2-Chloropropyldimethylammonium chloride

CAS Number: 4584-49-0
MDL Number: MFCD00012534
Molecular Formula: C5H12ClN
Molecular Weight: 158.07
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 189


Other Product Names / Synonyms: 2-(Dimethylamino)isopropyl Chloride Hydrochloride

Formula: C₅H₁₃Cl₂N
MW: 158.07 g/mol
Melting Pt: 189 °C
MDL Number: MFCD00012534
CAS Number: 4584-49-0
EINECS: 224-971-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7feb1aa2ffd7d478685acbc1639e2ce7a137b9ee&vid=7feb1aa2ffd7d478685acbc1639e2ce7a137b9ee&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR