3,5-Dimethyl-1H-pyrazole ≥98.0% (by GC, titration analysis)

Supplier: TCI America

D1647-500G D1647-25G D1647-100G
TCD1647-500GEA 215.34 CAD
TCD1647-500G TCD1647-025G TCD1647-100G
3,5-Dimethyl-1H-pyrazole ≥98.0% (by GC, titration analysis)
3,5-Dimethyl-1H-pyrazole

CAS Number: 67-51-6
MDL Number: MFCD00005243
Molecular Formula: C5H8N2
Molecular Weight: 96.13
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 218
Melting point (°C): 108

Formula: C₅H₈N₂
Boiling Pt: 218 °C
Melting Pt: 108 °C
Density: 0.8839 g/cm³ (16 °C)
MDL Number: MFCD00005243
CAS Number: 67-51-6
EINECS: 200-657-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=afb149664cbd74a0c14af1dbc4e2d6b00ac0cecb&vid=afb149664cbd74a0c14af1dbc4e2d6b00ac0cecb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR