trans-2-Decenal ≥93.0%

Supplier: TCI America

Synonyms: trans-2-Decenal, (E)-2-Decenal, (E)-Dec-2-enal

D1642-5ML D1642-25ML
TCD1642-005MLEA 72.68 CAD
TCD1642-005ML TCD1642-025ML
trans-2-Decenal ≥93.0%
trans-2-Decenal

CAS Number: 3913-81-3
MDL Number: MFCD00014679
Molecular Formula: C10H18O
Molecular Weight: 154.25
Purity/Analysis Method: >93.0% (GC)
Form: Clear Liquid
Boiling point (°C): 80
Flash Point (°C): 108
Specific Gravity (20/20): 0.85

Formula: C₁₀H₁₈O
MW: 154.25 g/mol
Boiling Pt: 80 °C
Density: 0.85 (20 °C)
Flash Pt: 108 °C
Storage Temperature: Ambient
MDL Number: MFCD00014679
CAS Number: 3913-81-3
EINECS: 223-474-2
UN: 3082
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=71a1672a5a1eef8c029b09d4c81fd7da3026740a&vid=c2756997c0c519b6391c06190132f727b0656cd8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 93.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR