ω,ω-Diethoxyacetophenone ≥95.0%

Supplier: TCI America

Synonyms: 2,2-Diethoxyacetophenone, 2,2-Diethoxy-1-phenylethan-1-one

D1640-500G D1640-25G
TCD1640-500GEA 543.52 CAD
TCD1640-500G TCD1640-025G
ω,ω-Diethoxyacetophenone ≥95.0%
ω,ω-Diethoxyacetophenone

CAS Number: 6175-45-7
MDL Number: MFCD00009659
Molecular Formula: C12H16O3
Molecular Weight: 208.26
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 134
Flash Point (°C): 127
Specific Gravity (20/20): 1.04


Product Categories: Materials Chemistry, Polymerization Initiators
Other Product Names / Synonyms: Benzoylformaldehyde Diethyl Acetal, Phenylglyoxal Diethyl Acetal

Formula: C₁₂H₁₆O₃
MW: 208.26 g/mol
Boiling Pt: 131
Density: 1.04 (20 °C)
Flash Pt: 127 °C
MDL Number: MFCD00009659
CAS Number: 6175-45-7
EINECS: 228-220-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fca07ed4947ece4afa36da1809151ffab79df8fb&vid=fca07ed4947ece4afa36da1809151ffab79df8fb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR