4,4'-Dichlorobenzophenone ≥99.0%

Supplier: TCI America

Synonyms: 4,4-Dichlorobenzophenone, 4,4-DBP, Bis(4-chlorophenyl) ketone

D1621-25G D1621-500G D1621-100G
TCD1621-025GEA 75.57 CAD
TCD1621-025G TCD1621-500G TCD1621-100G
4,4'-Dichlorobenzophenone ≥99.0%
4,4'-Dichlorobenzophenone

CAS Number: 90-98-2
MDL Number: MFCD00000623
Molecular Formula: C13H8Cl2O
Molecular Weight: 251.11
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 353
Melting point (°C): 147

Formula: C₁₃H₈Cl₂O
MW: 251.11 g/mol
Boiling Pt: 353 °C
Melting Pt: 147 °C
MDL Number: MFCD00000623
CAS Number: 90-98-2
EINECS: 202-030-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6b8c587f44e1b780293700e53691e0af2aaa0e6a&vid=6b8c587f44e1b780293700e53691e0af2aaa0e6a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR