Dicyclohexyl sulfide ≥90.0%

Supplier: TCI America

D1617-25ML D1617-500ML
TCD1617-025MLEA 31.6 CAD
TCD1617-025ML TCD1617-500ML
Dicyclohexyl sulfide ≥90.0%
Dicyclohexyl sulphide

CAS Number: 2550-40-5
MDL Number: MFCD00013759
Molecular Formula: C12H22S2
Molecular Weight: 230.43
Purity/Analysis Method: >90.0% (GC)
Form: Clear Liquid
Boiling point (°C): 110
Specific Gravity (20/20): 1.05


Product Categories: Synthetic Organic Chemistry, Sulfenylation
Other Product Names / Synonyms: Cyclohexyl Disulfide

Formula: C₁₂H₂₂S₂
Boiling Pt: 110 °C
Density: 1.05 (20 °C)
MDL Number: MFCD00013759
CAS Number: 2550-40-5
EINECS: 219-851-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d4f90b16c8b456ed6b4aeebdb3790e66d4a8ac2b&vid=d4f90b16c8b456ed6b4aeebdb3790e66d4a8ac2b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR