4,4'-Dimethoxytrityl chloride ≥97.0% (by HPLC, titration analysis)

Supplier: TCI America

D1612-25G D1612-5G
TCD1612-025GEA 194.34 CAD
TCD1612-025G TCD1612-005G
4,4'-Dimethoxytrityl chloride ≥97.0% (by HPLC, titration analysis)
4,4'-Dimethoxytrityl chloride

[Hydroxyl Protecting Agent]
CAS Number: 40615-36-9
MDL Number: MFCD00008409
Molecular Formula: C21H19ClO2
Molecular Weight: 338.83
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Crystal
Melting point (°C): 125


Product Categories: Synthetic Organic Chemistry, Protection & Derivatization Reagents (for Synthesis), Nucleic Acid Synthesis
Other Product Names / Synonyms: DMT-Cl

Formula: C₂₁H₁₉ClO₂
Melting Pt: 125 °C
MDL Number: MFCD00008409
CAS Number: 40615-36-9
EINECS: 255-002-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5cdadcea0ae0eb17deb1f3c4972ff06a82b49d88&vid=ae6b8c39712beb037b8b2ba7c34e0aff70bfd5bc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR