2,3-Dimethylindole ≥98.0%

Supplier: TCI America

D1579-250G D1579-25G
TCD1579-250GEA 598.82 CAD
TCD1579-250G TCD1579-025G
2,3-Dimethylindole ≥98.0%
2,3-Dimethylindole

CAS Number: 91-55-4
MDL Number: MFCD00005617
Molecular Formula: C10H11N
Molecular Weight: 145.21
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 285
Melting point (°C): 107

Formula: C₁₀H₁₁N
Boiling Pt: 285 °C
Melting Pt: 107 °C
MDL Number: MFCD00005617
CAS Number: 91-55-4
EINECS: 202-076-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ad42e568bee116dd0a32f244016d0d8256f36b8&vid=4ad42e568bee116dd0a32f244016d0d8256f36b8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR