5,6-Dimethyl-1,10-phenanthroline ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

D1559-100MG D1559-1G
TCD1559-100MGEA 120.71 CAD
TCD1559-100MG TCD1559-1G
5,6-Dimethyl-1,10-phenanthroline ≥98.0% (by HPLC, titration analysis)
5,6-Dimethyl-1,10-phenanthroline

CAS Number: 3002-81-1
MDL Number: MFCD00004983
Molecular Formula: C14H12N2
Molecular Weight: 208.26
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal

Formula: C₁₄H₁₂N₂
MDL Number: MFCD00004983
CAS Number: 3002-81-1
EINECS: 221-100-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=34e025764ab9784c820691b0112ace7369e9d141&vid=34e025764ab9784c820691b0112ace7369e9d141&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR