3,5-Dimethoxybenzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

D1534-500G D1534-25G
TCD1534-500GEA 462.78 CAD
TCD1534-500G TCD1534-025G
3,5-Dimethoxybenzoic acid ≥98.0% (by GC, titration analysis)
3,5-Dimethoxybenzoic acid

CAS Number: 1132-21-4
MDL Number: MFCD00002502
Molecular Formula: C9H10O4
Molecular Weight: 182.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 183

Formula: C₉H₁₀O₄
MW: 182.18 g/mol
Boiling Pt: 341 °C (1013 hPa)
Melting Pt: 183 °C
Density: 1.423 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002502
CAS Number: 1132-21-4
EINECS: 214-473-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab826852e912f363a10c38f38ccf3098f6b4945d&vid=8267fd7afe76a852699b35742c16f31b78eacb3a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR