Dibenzo-18-crown-6-(dibenzo-1,4,7,10,13,16-hexaoxacyclooctadecane, tin(II) ionophore I) ≥99.0%

Supplier: TCI America

D1533-5G D1533-25G
TCD1533-005GEA 26.86 CAD
TCD1533-005G TCD1533-025G
Dibenzo-18-crown-6-(dibenzo-1,4,7,10,13,16-hexaoxacyclooctadecane, tin(II) ionophore I) ≥99.0%
Dibenzo-18-crown-6 (Dibenzo-1,4,7,10,13,16-hexaoxacyclooctadecane, tin (II) ionophore I)

CAS Number: 14187-32-7
MDL Number: MFCD00005098
Molecular Formula: C20H24O6
Molecular Weight: 360.41
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 382
Melting point (°C): 163

Formula: C₂₀H₂₄O₆
Boiling Pt: 382 °C
Melting Pt: 163 °C
MDL Number: MFCD00005098
CAS Number: 14187-32-7
EINECS: 238-041-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=92b27f65e6ec14249bf5e3ccfebf1a20200d71a7&vid=92b27f65e6ec14249bf5e3ccfebf1a20200d71a7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR