2,5-Dimethylpyrazine ≥80.0% (contains 2,6-isomer)

Supplier: TCI America
Warning

D1526-25ML D1526-500ML
TCD1526-025MLEA 45.82 CAD
TCD1526-025ML TCD1526-500ML
2,5-Dimethylpyrazine ≥80.0% (contains 2,6-isomer)
2,5-Dimethylpyrazine

CAS Number: 123-32-0
MDL Number: MFCD00006147
Molecular Formula: C6H8N2
Molecular Weight: 108.14
Purity/Analysis Method: >80.0% (GC)
Form: Clear Liquid
Boiling point (°C): 182
Flash Point (°C): 64
Freezing point (°C): 15
Specific Gravity (20/20): 0.99

Formula: C₆H₈N₂
MW: 108.14 g/mol
Boiling Pt: 182 °C
Melting Pt: 15 °C
Density: 0.99 (20 °C)
Flash Pt: 64 °C
Storage Temperature: Ambient
MDL Number: MFCD00006147
CAS Number: 123-32-0
EINECS: 204-618-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e1794b70a3fbe5b387bad59cfc1e1c81a747bfb7&vid=e1794b70a3fbe5b387bad59cfc1e1c81a747bfb7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 80.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR