Dodecyltriethoxysilane ≥95.0%
Supplier: TCI America
Synonyms:
Triethoxy(dodecyl)silane
D1510-10ML
D1510-25ML
TCD1510-010MLEA
82.16
CAD
TCD1510-010ML
TCD1510-25ML
Dodecyltriethoxysilane ≥95.0%
Dodecyltriethoxysilane
CAS Number: 18536-91-9
MDL Number: MFCD00048437
Molecular Formula: C18H40O3Si
Molecular Weight: 332.60
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 129
Flash Point (°C): 110
Specific Gravity (20/20): 0.88
Product Categories: Materials Chemistry, Silane Coupling Agents
Other Product Names / Synonyms: Triethoxylaurylsilane
Formula:
C₁₈H₄₀O₃Si Boiling Pt: 129 °C Density: 0.88 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00048437 CAS Number: 18536-91-9 EINECS: 242-409-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...