Dansyl amide ≥98.0% (by titrimetric analysis)
Supplier: TCI America
D1507-5G
D1507-1G
TCD1507-005GEA
368.14
CAD
TCD1507-005G
TCD1507-001G
Dansyl amide ≥98.0% (by titrimetric analysis)
Dansyl amide
[for Fluorometry]
CAS Number: 1431-39-6
MDL Number: MFCD00004000
Molecular Formula: C12H14N2O2S
Molecular Weight: 250.32
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 218
Other Product Names / Synonyms: 5-Dimethylaminonaphthalene-1-sulfonamide
Formula:
C₁₂H₁₄N₂O₂S Boiling Pt: 405 °C (1013 hPa) Melting Pt: 218 °C |
MDL Number:
MFCD00004000 CAS Number: 1431-39-6 EINECS: 215-854-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...