4-(Dimethylamino)benzaldehyde ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
Ehrlich's Reagent, 4-(N,N-Dimethylamino)benzaldehyde, p-Dimethylamino-Benzaldehyde, p-Dimethylaminobenzaldehyde, p-(Dimethylamino)benzaldehyde
D1495-25G
D1495-500G
D1495-100G
TCD1495-025GEA
175.32
CAD
TCD1495-025G
TCD1495-500G
TCD1495-100G
4-(Dimethylamino)benzaldehyde ≥98.0% (by GC, titration analysis)
4-(Dimethylamino)benzaldehyde
CAS Number: 100-10-7
MDL Number: MFCD00003381
Molecular Formula: C9H11NO
Molecular Weight: 149.19
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 177
Melting point (°C): 75
Flash Point (°C): 164
Formula:
(CH₃)₂NC₆H₄CHO MW: 149.19 g/mol Boiling Pt: 176-177 Melting Pt: 72 Density: 1.0254 g/cm³ (99.9 °C) Flash Pt: 164 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003381 CAS Number: 100-10-7 EINECS: 202-819-0 Merck Index: 13,03257 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...