1,4-Dibromotetrafluorobenzene ≥98.0%

Supplier: TCI America
Warning

D1493-25G D1493-5G
TCD1493-025GEA 445.56 CAD
TCD1493-025G TCD1493-005G
1,4-Dibromotetrafluorobenzene ≥98.0%
1,4-Dibromotetrafluorobenzene

CAS Number: 344-03-6
MDL Number: MFCD00000309
Molecular Formula: C6Br2F4
Molecular Weight: 307.87
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 78

Formula: C₆Br₂F₄
MW: 307.87 g/mol
Boiling Pt: 198 °C (1013 hPa)
Melting Pt: 78 °C
MDL Number: MFCD00000309
CAS Number: 344-03-6
EINECS: 206-448-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bb0a16290283aceab2a43a34ed3c3571ff253ca3&vid=7df0d409f52ed59e38bde7e7b846779b0a4692ff&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR