3,5-Dimethoxyaniline ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

D1484-5G D1484-25G
TCD1484-005GEA 90.06 CAD
TCD1484-005G TCD1484-025G
3,5-Dimethoxyaniline ≥98.0% (by GC, titration analysis)
3,5-Dimethoxyaniline

CAS Number: 10272-07-8
MDL Number: MFCD00008392
Molecular Formula: C8H11NO2
Molecular Weight: 153.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 178
Melting point (°C): 57
Flash Point (°C): 110
Storage Temperature: 0-10°C

Formula: C₈H₁₁NO₂
Boiling Pt: 178 °C
Melting Pt: 57 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00008392
CAS Number: 10272-07-8
EINECS: 233-616-5
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1258f0a4977b69541bea9321f45cb2a8b9267405&vid=e407208b9699cfa34bbceb29c5170bdc4943ad92&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR