2,3-Dihydroxybenzaldehyde ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

D1478-5G D1478-25G
TCD1478-005GEA 135.88 CAD
TCD1478-005G TCD1478-025G
2,3-Dihydroxybenzaldehyde ≥98.0% (by GC, titration analysis)
2,3-Dihydroxybenzaldehyde

CAS Number: 24677-78-9
MDL Number: MFCD00003324
Molecular Formula: C7H6O3
Molecular Weight: 138.12
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 120
Melting point (°C): 107

Formula: C₇H₆O₃
Boiling Pt: 120 °C
Melting Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00003324
CAS Number: 24677-78-9
EINECS: 246-398-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=db951c6c70328cbffe6c04f6b9de71e463e43922&vid=5ee894983054850ab1642b9af7dfca819895cda8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR