3,5-Dichloroanthranilic acid ≥97.0% (by HPLC, titration analysis)

Supplier: TCI America

D1475-25G D1475-5G
TCD1475-025GEA 271.13 CAD
TCD1475-025G TCD1475-005G
3,5-Dichloroanthranilic acid ≥97.0% (by HPLC, titration analysis)
3,5-Dichloroanthranilic acid

CAS Number: 2789-92-6
MDL Number: MFCD00017088
Molecular Formula: C7H5Cl2NO2
Molecular Weight: 206.02
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Crystal
Melting point (°C): 233


Other Product Names / Synonyms: 2-Amino-3,5-dichlorobenzoic Acid

Formula: C₇H₅Cl₂NO₂
Boiling Pt: 344 °C (1013 hPa)
Melting Pt: 233 °C
MDL Number: MFCD00017088
CAS Number: 2789-92-6
EINECS: 220-520-3
UN: 2811
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8eabc52da3d56e9ada04365c27e15a4ec24ed701&vid=0aa7089fc4072bea129f97b23c8581f8ace84fc5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR