(2,5-Dimethoxyphenyl)acetic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

D1445-5G D1445-25G
TCD1445-005GEA 80.58 CAD
TCD1445-005G TCD1445-025G
(2,5-Dimethoxyphenyl)acetic acid ≥98.0% (by GC, titration analysis)
(2,5-Dimethoxyphenyl)acetic acid

CAS Number: 1758-25-4
MDL Number: MFCD00004322
Molecular Formula: C10H12O4
Molecular Weight: 196.20
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 124

Formula: (C₆H₃(OCH₃)₂)CH₂COOH
Boiling Pt: 327 °C (1013 hPa)
Melting Pt: 124 °C
MDL Number: MFCD00004322
CAS Number: 1758-25-4
EINECS: 217-151-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c9626f3245c4e6aac999b2cee0ab8f6560cc0800&vid=c9626f3245c4e6aac999b2cee0ab8f6560cc0800&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR