2',5'-Dimethoxyacetophenone ≥98.0%

Supplier: TCI America

D1444-25G D1444-500G
TCD1444-025GEA 86.9 CAD
TCD1444-025G TCD1444-500G
2',5'-Dimethoxyacetophenone ≥98.0%
2',5'-Dimethoxyacetophenone

CAS Number: 1201-38-3
MDL Number: MFCD00008728
Molecular Formula: C10H12O3
Molecular Weight: 180.20
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 158
Flash Point (°C): 113
Freezing point (°C): 18
Specific Gravity (20/20): 1.14

Formula: C₁₀H₁₂O₃
Boiling Pt: 158 °C
Melting Pt: 18…20 °C
Density: 1.14 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00008728
CAS Number: 1201-38-3
EINECS: 214-858-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1a258c277083fdecf9c18a95ebbd41056798fc00&vid=1a258c277083fdecf9c18a95ebbd41056798fc00&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR